Learn More
Marienfeld Superior Counting chambers, DIN 12847 compliant, Thoma new, depth 0.1 mm, double netting, bright lines,
Brand:  Marienfeld Superior 0640830
Counting chambers, DIN 12847 compliant, Thoma new, depth 0.1 mm, double netting, bright lines,
Product Code. 17885894
  • 81.82€ / Each
Only null left
Estimated Shipment: 24-02-2026