Learn More
Marienfeld Superior Counting chambers, DIN 12847 compliant, Thoma new, depth 0.1 mm, double netting, bright lines,
Brand: Marienfeld Superior 0640830
Counting chambers, DIN 12847 compliant, Thoma new, depth 0.1 mm, double netting, bright lines,