Learn More
Marienfeld Superior™ Counting Chamber with Bright Line Double Net Ruling
Product Code. 15827045
Change view
Click to view available options
Grid Type:
Burker
Neubauer
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Grid Type Unit Size
15827045 Burker Each
15748128 Neubauer Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15827045 Supplier Marienfeld Superior™ Supplier No. 0640231
  • ONLINE EXCLUSIVE PRICE
    104.16€
    76.99€ / Pack of 1
    Save 27.17€
    26% Off
 Restricted Item
Estimated Shipment: 21-12-2026
Marienfeld Superior™ Counting Chamber with double net ruling are precise measuring instruments for investigation of cell suspensions
  • Precise measuring instrument for determination of the number of particles in liquids
  • Made of special optical glass
  • Equipped with two grids
  • With 2 haemacytometer cover glasses 20x26 mm
  • Individually packed in a transparent plastic box, 10 boxes in a carton
Specifications
Product Type Counting Chamber, Bright line
Grid Type Burker
Description CE-marked (IVD)
Cell Size 0.0025 mm2
Material Special optical glass
Quantity 1
Depth (Metric) 0.1 mm
Includes Two cover glasses 20 x 26 mm and two clamps