Learn More
Hecht Karl™ Counting Chamber
Product Code. 10647564
Change view
Click to view available options
Grid Type:
16 large 1 mm sq. squares with each square divided into 16 smaller squares each with a side length of 0.25 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Grid Type Unit Size
10647564 16 large 1 mm sq. squares with each square divided into 16 smaller squares each with a side length of 0.25 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10647564 Supplier Hecht Karl™ Supplier No. 40449712
  • ONLINE EXCLUSIVE PRICE
    118.53€
    69.05€ / Pack of 1
    Save 49.48€
    41% Off
Estimated Shipment: 28-12-2026
Specifications
Product Type Fuchs-Rosenthal Counting Chamber
Grid Type 16 large 1 mm sq. squares with each square divided into 16 smaller squares each with a side length of 0.25 mm
For Use With (Application) Commonly used to count cells in cerebrospinal fluid