Learn More
Coumaphos-d10, TRC
Specifications
Specifications
| Chemical Name or Material | Coumaphos-d10 |
| CAS | 287397-86-8 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C14 2H10 H6 Cl O5 P S |
| Quantity | 1 mg |
| Synonym | Coumaphos D10 (di(ethyl D5)) |
| InChI Formula | InChI=1S/C14H16ClO5PS/c1-4-17-21(22,18-5-2)20-10-6-7-11-9(3)13(15)14(16)19-12(11)8-10/h6-8H,4-5H2,1-3H3/i1D3,2D3,4D2,5D2 |
| SMILES | [2H]C([2H])([2H])C([2H])([2H])OP(=S)(Oc1ccc2C(=C(Cl)C(=O)Oc2c1)C)OC([2H])([2H])C([2H])([2H])[2H] |
| IUPAC Name | 7-[bis(1,1,2,2,2-pentadeuterioethoxy)phosphinothioyloxy]-3-chloro-4-methylchromen-2-one |
| Molecular Weight (g/mol) | 372.83 |
| Show More |