Learn More
Sartorius™ Cotton Linters FN 6 Chromatography Paper
Product Code. 10307841 Shop All Sartorius Products
Click to view available options
Length (Metric):
580 mm
Unit Size:
Pack of 100
Product Code. 10307841
Brand: Sartorius™ FT2506580600N
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Specifications
Format Sheet
Grade FN 6
Material Cotton Linters
Width (Metric) 600 mm
Length (Metric) 580 mm
Technique Chromatography