Learn More
Sartorius™ Cotton Linters FN 3 Chromatography Paper
Product Code. 10400282 Shop All Sartorius Products
Click to view available options
Length (Metric):
250 mm
300 mm
580 mm
3460 mm
Width (Metric):
250 mm
570 mm
580 mm
600 mm
Unit Size:
Pack of 100
Product Code. 10400282
Brand: Sartorius™ FT2503300580N
Specifications
Format Sheet
Grade FN 3
Material Cotton Linters
Width (Metric) 580 mm
Length (Metric) 300 mm
Technique Chromatography