Learn More
Corning™ Costar™ Stripettor™ Pipet Filler/Dispenser Replacement Parts
GREENER_CHOICE
Product Code. p-3573267 Shop All Corning Products
Click to view available options
Type:
Pipet Holder
Recharger/Adapter
Replacement Filter, 0.2μm
Replacement Filter, 0.45μm
Unit Size:
Each
Pack of 5
Product Code. 11901991
Brand: Corning™ 4914
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
For use with Stripettor™ filler/dispenser
  • PTFE filter (pack of 5) is for the Stripettor™ Plus pipetting controller
  • Pore Size: 0.45 μm
  • Autoclavable
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Not Autoclavable
Type Recharger/Adapter
For Use With (Equipment) Costar™ stripettor pipet filler/dispenser
Electrical Requirements 120V 60Hz
Voltage 120 V
Frequency 60 Hz