Learn More
Corning™ Snapwell™ Culture Inserts
GREENER_CHOICE
Product Code. 10670692 Shop All Corning Products
Click to view available options
Material:
PET
Polycarbonate
Pore Density:
1 x 108 pores/cm2
4 x 106 pores/cm2
Unit Size:
Pack of 24
You may only order up to the available stock. For more details call customer services on +351 21 425 33 50.
Product Code. 10670692
Brand: Corning™ 3801
You may only order up to the available stock. For more details call customer services on +351 21 425 33 50.
Designed with a ring supported membrane for use with diffusion or Ussing chambers once cells are grown to confluence. Corning™ Snapwell™ Culture Inserts are primarily used for transport and electrophysiological studies.
The Snapwell insert is a modified Transwell culture insert that contains a 12 mm diameter tissue culture-treated membrane supported by a detachable ring. The inserts are primarily used for transport and electrophysiological studies. Once cells are grown to confluence, this ring-supported membrane can be placed into either vertical or horizontal diffusion or Ussing chambers.
Order Info
Sterile culture inserts are packaged in 6-well cluster plates.
TRUSTED_SUSTAINABILITY
Specifications
Description 6 inserts and 6-well plate
Culture Area 1.12 cm2
Material PET
Pore Density 4 x 106 pores/cm2
Pore Size 0.4 μm
Type Culture Insert
Diameter (Metric) Outer 12 mm
No. per Pack 6/Pk.
No. per Case 24/Cs.