Learn More
Corning™ Costar™ Snap Cap Microcentrifuge Tubes
Product Code. 10439521 Shop All Corning Products
Click to view available options
Color:
Assorted
Natural
Capacity (Metric):
0.65 mL
1.7 mL
2 mL
Packaging:
1000/Pk.
500/Pk.
5000/Pk.
Unit Size:
Pack of 1000
Pack of 500
Pack of 5000
Product Code. 10439521
Brand: Corning™ 3621
Features a snap cap with a positive seal that allows for repeated opening and closing with flat-cap surfaces for convenient labeling. Corning™ Costar™ Snap Cap Microcentrifuge Tubes are compatible with leading microcentrifuge rotors.
  • Supplied nonsterile and are autoclavable.
  • External graduations and frosted writing spot for easy sample identification.
  • Positive seal design allows repeated opening and closing.
  • Flat cap surface for convenient labeling.
  • Withstands a maximum RCF of 17,000 x g.
  • RNase-/DNase-free.
TRUSTED_SUSTAINABILITY
Specifications
Cap Style Snap Cap
Certifications/Compliance RNase, DNase-free
Closure Color Natural
Closure Material Polypropylene
Autoclavable Autoclavable
Capacity (Metric) 1.7 mL
Color Natural
Material Polypropylene
No. per Pack 500/Pk.
Packaging 5000/Pk.
Sterility Non-sterile
Graduated Yes
Closure Type Snap Cap
Bottom Shape Conical
Max. RCF 17,000 x g
Product Type Microcentrifuge Tube