Learn More
Corning™ Thermowell™ 96-Well Polypropylene PCR Microplates
Product Code. 10727811 Shop All Corning Products
Change view
Click to view available options
Color:
Clear
Natural
Barcode:
Includes Barcode
No Barcode
Unit Size:
Pack of 25
Pack of 50
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Barcode unitSize
10727811 Natural No Barcode Pack of 25
10363173 Clear Includes Barcode Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10727811 Supplier Corning™ Supplier No. 6551
Only null left
Add to basket
Add to basket
Constructed of medical grade polypropylene to accommodate wide temperature changes that might occur from storage to thermal cycling. Corning™ Thermowell™ 96-Well Polypropylene PCR Microplates are ideal for the polyerase chain reaction (PCR) method of DNA amplification.
Corning Thermowell GOLD 96-well PCR microplates are offered in four formats to ensure maximum flexibility and a perfect match for your applications. The original Thermowell 96-well PCR microplates are universal fit and can be cut into 3 x 8 well segments.
TRUSTED_SUSTAINABILITY
Specifications
Barcode No Barcode
Certified DNase and RNase-free
Format Standard
Lid Without Lid
Description Can be cut into 3 x 8 well strips for lower throughput applications. Available PCR plate adapter for use with ABI PRISM™ 3700 DNA Analyzer
Array 8 x 12
Color Natural
Material Polypropylene
No. of Wells 96
Sterility Non-sterile
Total Volume (Metric) Well 200 μL
Well Shape Conical Bottom (V-well)
For Use With (Application) PCR
No. per Case 25/Cs.
No. per Pack 25/Pk.
Quantity 25/Pk.