Learn More
Isofield™ Cos Non-Sterile Lightweight Cleanroom Sock - GMP Annex 1 Compatible
BIOPHARMA
Product Code. 30178852
Change view
Click to view available options
Size:
Large/X-Large
Small/Medium
Unit Size:
500 pairs
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
30178852 Large/X-Large 500 pairs
30178833 Small/Medium 500 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30178852 Supplier Isofield™ Supplier No. 70610LXL
Safety Starts Here: Save Up to 60%
Use promo code 28008
View all promotions
Estimated Shipment: 09-12-2026
The Isofield Cos single-use non-sterile lightweight cleanroom sock is GMP Annex 1 compatible, offering comfort and protection in cleanroom settings. Available in three sizes, the Cos’ hourglass heel design ensures comfort levels rivalling most reusable alternatives.
  • Non-sterile lightweight
  • Cleanroom socks
  • Single-use
  • 95% polyester, 5% spandex
  • Mid-calf length
  • Low linting
  • White color
Specifications
Size Large/X-Large
Product Type Cleanroom Socks