missing translation for 'onlineSavingsMsg'
Learn More

Cortisone, TRC

Product Code. 17954835
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17954835 25 g 25g
17911925 5 g 5g
17963295 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17954835 Supplier Toronto Research Chemicals Supplier No. TRCC69650025G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53-06-5
Molecular Formula C21 H28 O5
Molecular Weight (g/mol) 360.44
Synonym Cortisone (8CI), 17,21-Dihydroxypregn-4-ene-3,11,20-trione, 11-Dehydro-17-hydroxycorticosterone, 17-Hydroxy-11-dehydrocorticosterone, 17α,21-Dihydroxy-4-pregnene-3,11,20-trione, 17α,21β-Dihydroxy-4-pregnene-3,11,20-trione, 17α-Hydroxy-11-dehydrocorticosterone, Adrenalex, Compound E, Cortisate, Cortivite, Cortogen, Cortone, KE, Kendall's compound E, NSC 9703, Pregn-4-en-17α,21-diol-3,11,20-trione, Reichstein's substance Fa, Wintersteiner's compound F, δ4-Pregnene-17α,21-diol-3,11,20-trione
IUPAC Name (8S,9S,10R,13S,14S,17R)-17-hydroxy-17-(2-hydroxyacetyl)-10,13-dimethyl-1,2,6,7,8,9,12,14,15,16-decahydrocyclopenta[a]phenanthrene-3,11-dione
SMILES C[C@]12CCC(=O)C=C1CC[C@H]3[C@@H]4CC[C@](O)(C(=O)CO)[C@@]4(C)CC(=O)[C@H]23

Specifications

Quantity 25 g
InChI Formula InChI=1S/C21H28O5/c1-19-7-5-13(23)9-12(19)3-4-14-15-6-8-21(26,17(25)11-22)20(15,2)10-16(24)18(14)19/h9,14-15,18,22,26H,3-8,10-11H2,1-2H3/t14-,15-,18+,19-,20-,21-/m0/s1
Formula Weight 360.1937
Percent Purity >95%
Chemical Name or Material Cortisone
Grade HPLC
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.