Learn More
Corning™ Thermowell™ Gold PCR Cap Strips
GREENER_CHOICE
Product Code. 10531673 Shop All Corning Products
Change view
Click to view available options
Color:
Assorted
Clear
Product Type:
Dome Cap Strip
Flat Cap Strip
Unit Size:
Pack of 1250
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Product Type unitSize
10531673 Assorted Dome Cap Strip Pack of 1250
10571493 Clear Flat Cap Strip Pack of 1250
10307722 Clear Dome Cap Strip Pack of 1250
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10531673 Supplier Corning™ Supplier No. 3748
Only null left
Add to basket
Add to basket
Fit all Thermowell Gold 1 x 8-well PCR strips and 96-well PCR microplates
Tube strips consist of eight 0.2 mL thin wall polypropylene tubes connected together. Dual connectors between adjacent tubes eliminate inadvertent breakage during sample handling. Tube strips are designed for precise fit in thermal cyclers to optimize heat transfer. Thermowell GOLD cap strips are sold separately from Thermowell GOLD tube strips. Original Thermowell tube strips and cap strips are packaged together. Tube strips are RNase-/DNase-free, and are autoclavable at 121°C.
Cat. No. 6547 has 60 of each color per bag; 1 bag of each color per case.
Cat. No. 3742 is designed for real-time PCR. Suitable for use with Thermowell GOLD 0.2 mL 1 x 8 PCR tube strips and 96-well microplates.
TRUSTED_SUSTAINABILITY
Specifications
Color Assorted
Material Polypropylene
Sterility Non-sterile
Autoclavable Autoclavable
No. per Case 1250/Cs.
No. per Pack 125/Pk.
For Use With (Application) PCR
Product Type Dome Cap Strip
Plate Compatibility Thermowell Gold 1 x 8-well PCR Strips and 96-well PCR Microplates