Learn More
Corning™ Thermowell™ Gold PCR Cap Strips
Shop All Corning Products
Click to view available options
Color:
Assorted
Clear
Product Type:
Dome Cap Strip
Flat Cap Strip
Unit Size:
Pack of 1250
Description
Tube strips consist of eight 0.2 mL thin wall polypropylene tubes connected together. Dual connectors between adjacent tubes eliminate inadvertent breakage during sample handling. Tube strips are designed for precise fit in thermal cyclers to optimize heat transfer. Thermowell GOLD cap strips are sold separately from Thermowell GOLD tube strips. Original Thermowell tube strips and cap strips are packaged together. Tube strips are RNase-/DNase-free, and are autoclavable at 121°C.
Cat. No. 6547 has 60 of each color per bag; 1 bag of each color per case.
Cat. No. 3742 is designed for real-time PCR. Suitable for use with Thermowell GOLD 0.2 mL 1 x 8 PCR tube strips and 96-well microplates.
Spécification
Spécification
| Color | Clear |
| Material | Polypropylene |
| Sterility | Non-sterile |
| Autoclavable | Autoclavable |
| No. per Case | 1250/Cs. |
| No. per Pack | 125/Pk. |
| For Use With (Application) | PCR |
| Product Type | Dome Cap Strip |
| Plate Compatibility | Thermowell Gold 1 x 8-well PCR Strips and 96-well PCR Microplates |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu