Description
Corning Thermowell GOLD 384-well PCR microplates feature exceptional dimensional stability following thermal cycling, and are fully compatible with automation, commonly used thermal cyclers, and Applied Biosystems™ sequencing adapters (see compatibility table).
Order Info
Packaged in packs of 10.
Certifications
Certified DNase- and RNase-free.
Specifications
Specifications
| Format | Standard |
| Lid | Without Lid |
| Array | 16 x 24 |
| Color | Clear |
| Material | Polypropylene |
| No. of Wells | 384 |
| Sterility | Non-sterile |
| Well Shape | Round, Conical Bottom (V-well) |
| For Use With (Application) | For automation |
| No. per Case | 50/Cs. |
| Show More |