Learn More
Corning™ Thermowell™ 96-Well Polycarbonate PCR Microplates
GREENER_CHOICE
Product Code. 10130961 Shop All Corning Products
Change view
Click to view available options
Product Type:
PCR Microplate, Model M
Unit Size:
Pack of 25
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type unitSize
10130961 PCR Microplate, Model M Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10130961 Supplier Corning™ Supplier No. 6511
Only null left
Add to basket
Add to basket
Manufactured from ultrathin polycarbonate for fast, consistent heat transfer. Corning™ Thermowell™ 96-Well Polycarbonate PCR Microplates are ideal for the polyerase chain reaction (PCR) method of DNA amplification.
  • Low protein-binding surface prevents loss of sample
  • For oil-free operation, use polyethylene or aluminum sealing tape in conjunction with heated-lid thermal cyclers; also accommodates silicone reusable sealing mat
  • Withstands temperatures to 135°C without warping
  • Various well shapes fit popular thermal cyclers
  • Surface of DNA-BIND™ plate allows covalent attachment of ssDNA at 5« and 3« end
  • Nonsterile
Certifications
Polypropylene plates are certified DNase- and RNase-free.
TRUSTED_SUSTAINABILITY
Specifications
Format Standard
Lid Without Lid
Array 8 x 12
Binding Type Medium Binding
Color Clear
Product Type PCR Microplate, Model M
For Use With (Equipment) MJ Research PTC-100-96V, PTC-200 DNA EngineTM, Biometra Uno - ThermoblockerTM,Coy Corporation Temp Cycler II, Corbett Research FTS-960, Hybaid OmniGene withMicroblock, Quatro BioSystems T-C-40
Material Polycarbonate
No. of Wells 96
Sterility Non-sterile
Well Shape Round
For Use With (Application) PCR
No. per Case 25/Cs.
No. per Pack 1/Pk.
Well Color Clear