Learn More
Corning™ Synthemax™ II-SC Substrate
Product Code. 15319397 Shop All Corning Products
Change view
Click to view available options
Quantity:
10 mg (vial)
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15319397 10 mg (vial) 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15319397 Supplier Corning™ Supplier No. 3535
 Restricted Item
Estimated Shipment: 24-11-2026
Self-coating synthetic surface for stem cells
Easy-to-use, self-coatingmaterial for creating a unique synthetic surface that mimics the natural cell environment. Surface supportsthe adhesion and expansion of stem cells in their undifferentiated state and directed differentiation into specialized cell types. In addition, it offers a synthetic, xeno-free alternative to biological coatings and/or feeder cell layers used in hES and iPS cell culture.
Allows for the scalable multi-passage expansion of pluripotent stem cells in defined media with:
  • Stable doubling time
  • Phenotypicmarker expression (Oct4, SSEA4)
  • Retention of pluripotency
  • Retention of normal karyotype
Corning Synthemax II-SC Substrate is:
  • Easy-to-use
  • Able to coat plastic and glass surfaces
  • Lot-traceable
  • Gamma sterilized
  • Quality tested for lot-to-lot consistency
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Creating a unique synthetic surface that mimics the natural cell environment
Quantity 10 mg (vial)
Substrate Type Synthemax™ II-SC Substrate
Includes 1 mg/mL Synthemax II SC stock solution, sterile water, 0.025 mg/mL final volume of Corning™ Synthemax II SC working solution
Storage Requirements -20°C
Substrate Quantity 10 mg