Learn More
Corning™ Clear Polystyrene High Bind Stripwell™ Strips, without Frame or Lid, Non-sterile
Product Code. 10226771 Shop All Corning Products
Change view
Click to view available options
Surface Treatment:
High Binding
Unit Size:
Pack of 800
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Surface Treatment Unit Size
10226771 High Binding Pack of 800
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10226771 Supplier Corning™ Supplier No. 2580
  • ONLINE EXCLUSIVE PRICE
    545.00€
    498.13€ / Pack of 800
    Save 46.87€
    8% Off
Estimated Shipment: 09-04-2027
Designed and treated to provide uniform binding, high optical clarity, and low background interference. Corning™ Stripwell™ Strips provide a superior solution for in vitro diagnostic assays.
Corning™ Stripwell™ microplates may be used in a variety of laboratory assays. The flat bottom strips are designed to easily break apart and are pre-assembled in an “egg-crate” style strip holder that allows each individual well to be positioned back into the microplate once broken.
  • Stripwell microplates have 96-well flat bottom polystyrene format.
  • Low volume and standard Stripwell microplates have well volumes of 190 μL and 360 μL, respectively.
  • 1 x 8 strips are designed to fit only one way into the strip holder, eliminating the chance of misorientation.
TRUSTED_SUSTAINABILITY
Specifications
Format Strip Plate
Lid Without Lid
Array 8 x 12
Binding Type High Bind
Color Clear
Material Polystyrene
No. of Wells 96
Volume (Metric) Working 75 to 200 μL
Sterility Non-sterile
Surface Treatment High Binding
Total Volume (Metric) Well 360 μL
Well Shape Round, Flat Bottom (F-well)
For Use With (Application) In-vitro diagnostic assays
No. per Case 800/Cs.
No. per Pack 200/Pk.
Quantity 800/Cs.