Learn More
Corning™ Phenolic GPI Screw Caps for Disposable Culture Tubes
Click to view available options
Closure Size:
13-415
15-415
18-415
Height (Metric):
13.5 mm
16 mm
17.5 mm
Unit Size:
Pack of 1000
Pack of 250
Descrizione
Made of a phenolic compound. These caps have a white rubber liner.
Specifications
Specifications
| Closure Size | 18-415 |
| Liner Material | Rubber |
| Product Type | Screw Cap |
| Autoclavable | Autoclavable |
| Material | Phenolic |
| Color | Black |
| Height (Metric) | 17.5 mm |
| No. per Pack | 1000/Pk. |
| No. per Case | 1000/Cs. |
| Tip Material | Rubber |
| Show More |