Learn More
Corning™ Robolids with Corner Notch
Product Code. 10702973 Shop All Corning Products
Change view
Click to view available options
Format:
384-Well Robolid
96-Well Robolid
Unit Size:
Pack of 50
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Format unitSize
10702973 384-Well Robolid Pack of 50
10702783 96-Well Robolid Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10702973 Supplier Corning™ Supplier No. 3089
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
For use with automation equipment
  • Combines the sealing ability of a storage mat with the rigidity of a plastic lid
  • Designed for repeated application and removal by automation and for preventing short-term evaporation
  • Silicone sealing plugs for organic solvent resistance and low extractables
  • Can be used manually or with automation
  • Order Info
    Available in 96- and 384-well formats.
    TRUSTED_SUSTAINABILITY
    Specifications
    Sterility Non-sterile
    No. of Wells 384
    No. per Case 50/Cs.
    No. per Pack 25/Pk.
    Format 384-Well Robolid