Learn More
Corning™ Reusable Plastic Erlenmeyer Flasks with Red Graduations
Product Code. 17293544 Shop All Corning Products
Click to view available options
Capacity (Metric):
50 mL
100 mL
250 mL
500 mL
1 L
Graduations:
10 mL
20 mL
50 mL
100 mL
200 mL
Unit Size:
Pack of 4
Pack of 6
Product Code. 17293544
Brand: Corning™ 4990P100
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Translucent polymethylpentene; with polypropylene screw cap
Translucent polymethylpentene flasks are manufactured with red graduations for reference. The flasks have a screw neck and are supplied with a PP cap.
TRUSTED_SUSTAINABILITY
Specifications
Closure Material Polypropylene
Capacity (Metric) 100 mL
Material Polypropylene
Graduations 20 mL
Neck Style Narrow Mouth
No. per Case 6/Cs.
No. per Pack 1/Pk.
Closure Type Screw Cap