Learn More
Corning™ PureCoat™ Collagen Peptide Cell Culture Flasks with Vented Cap
Description
Corning ECM mimetic surfaces and substrates contain biologically active, synthetic, animal-free peptides that have been rationally designed to mimic the cell attachment motifs of native ECM proteins. The peptides enable optimal cell binding and signaling in a broad range of serum-free, xeno-free, and animal-free media formulations, supporting the propagation and differentiation of a range of stem, progenitor, and primary cell types.
Corning PureCoat cultureware is coated with biologically active, synthetic, animal-free peptides that are covalently linked to a proprietary surface to provide a highly consistent, cost-effective alternative to self-coated extracellular peptides. The proprietary covalent linkage orients the peptides for optimal binding and signaling.
Specifications
Specifications
| Capacity (Metric) | 175 cm2 |
| Material | Polystyrene |
| Neck Style | Canted |
| No. per Case | 10/Cs. |
| No. per Pack | 5/Pk. |
| Shape | Rectangular |
| Surface Coating | PureCoat E cm Mimetic Collagen I Peptide |
| Bottom Shape | Rectangular |
| Closure Type | Vented |
| Culture Area | 175 cm2 |