Learn More
Cole-Parmer™ Corning Polystyrene Spatula
Shop All Cole-Parmer Products
Click to view available options
Length (Metric):
246.06 mm
Unit Size:
Pack of 100
Specifications
Specifications
| Quantity | 100 Pk. |
| Sterility | Sterile |
| Shape | Spoon End, Round End |
| Certifications/Compliance | Raze/DNase Free |
| Disposable | Single-use |
| Product Type | Spatula |
| For Use With (Application) | For Research Labs That Require Contamination Free Sampling |
| Material | Polystyrene |
| Length (Metric) | 246.06 mm |