Learn More
Corning™ 384-Well, Cell Culture-Treated, Flat-Bottom Microplate
GREENER_CHOICE
Product Code. 10477221 Shop All Corning Products
Click to view available options
Color:
Clear
Unit Size:
Pack of 100
Product Code. 10477221
Brand: Corning™ 3701
Color:Clear; Sterility:Sterile; Lid:With Lid
  • Total well volume of 112 μL; working well volume of 20 μL – 80 μL
  • Cell culture microplates are sterile and nonpyrogenic.
  • The 384-well Universal Optics NBS microplate is manufactured using an advanced polymer with high clarity and improved chemical resistant properties.
  • TRUSTED_SUSTAINABILITY
    Specifications
    Format Standard
    Lid With Lid
    Culture Area 0.06 cm2
    Array 16 x 24
    Packaging 20/Pk., 100/Cs.
    Color Clear
    Material Polystyrene
    No. of Wells 384
    Volume (Metric) Working 20 to 80 μL
    Sterility Sterile
    Surface Treatment Cell Culture
    Well Shape Flat Bottom (F-well)
    For Use With (Application) Attachment and growth of anchorage-dependent cells
    No. per Case 100/Cs.
    No. per Pack 20/Pk.
    Quantity 100/Cs.