Learn More
Corning™ Plain Reusable Short Stem Plastic Funnels
Shop All Corning Products
Click to view available options
Diameter (Metric) Stem:
1.5 mm
4 mm
6 mm
7 mm
11 mm
15 mm
Height (Metric):
45 mm
65 mm
85 mm
110 mm
180 mm
220 mm
Length (Metric) Stem:
25 mm
35 mm
43 mm
55 mm
90 mm
95 mm
Unit Size:
Pack of 12
Pack of 24
Description
Allows safe and easy transfer of solids or liquids from one vessel to another.
Specifications
Specifications
| Diameter (Metric) Stem | 7 mm |
| Material | Polypropylene |
| Height (Metric) | 85 mm |
| Description | Constructed of durable translucent Polypropylene |
| Length (Metric) Stem | 43 mm |