Learn More
Corning™ Universal Carousel Rack for Six Lambda Plus Single-channel Pipettors
Shop All Corning Products
Description
This stand is designed to hold six Corning™ LambdaTM plus single-channel pipettors.
Specifications
Specifications
| Number of Channels | 1 |
| Product Line | Lambda Plus |
| Type | Carousel Rack |
| For Use With (Equipment) | 6 Pipettes |