Learn More
Corning™ Filtered IsoTip™ Universal Fit Racked Pipet Tips
Ideal for applications where avoiding cross-contamination is critical, such as DNA amplification and radioisotope handling
Brand:  Corning™ 4807
Additional Details : Weight : 1.22470kg
 View more versions of this product
Product Code. 10035510
  • 146.19€ / Pack of 960
Estimated Shipment: 13-12-2024
Description
  • IsoTip filtered pipet tips feature an inert, hydrophobic barrier that prevents aerosolized contaminants from coming in contact with pipettor shafts. (A Pipet Tip Compatibility Guide can be requested or downloaded from the Corning website.)
  • Ideal for applications where avoiding cross contamination is critical, such as DNA amplification and radioisotope handling.
  • Packaged sterile.
  • RNase-/DNase-free.
  • Nonpyrogenic.
Specifications
Specifications
0.2 to 10 μL
Sterile
Rack
Rack
Isotip
Filtered
Natural
Round
Gilson™ Pipetman™, Universal Fit Ultramicro