Learn More
Corning™ Epidermal Growth Factor (EGF), Mouse Natural (Receptor Grade)
Product Code. 11573520 Shop All Corning Products
Change view
Click to view available options
Quantity:
1 μg
100 μg
Purity or Quality Grade:
>95% by SDS-PAGE and >80% by pH 9.9 PAGE
>98% by SDS-PAGE and pH 9.9 PAGE
Unit Size:
100µg
1mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Purity or Quality Grade Unit Size
11573520 100 μg >98% by SDS-PAGE and pH 9.9 PAGE 100µg
11593600 1 μg >95% by SDS-PAGE and >80% by pH 9.9 PAGE 1mg
11533520 100 μg >95% by SDS-PAGE and >80% by pH 9.9 PAGE 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11573520 Supplier Corning™ Supplier No. 354010
 Restricted Item
Estimated Shipment: 11-03-2027
Enhance the proliferation of a wide variety of cell types
The Corning portfolio of cytokines, growth factors, and media supplementation offers a wide array of proteins that result in proliferation or differentiation of the affected cells.
  • Vialed, highly purified growth factors with high biological activity
  • Broad range of highly purified natural and recombinant lymphokines
  • Enriched culture supplements to be reconstituted/added in the medium of your choice
  • Propagate your cells under more defined serum-reduced or serum-free conditions
Growth factors are proteins that bind to receptors found on the surface of non-hematopoietic cells. Each family of growth factors affect specific cell types, e.g., nerve growth factors (NGF) affect nerve cell types, and epidermal growth factors (EGF) affect epithelial cell types.
Corning offers a range of growth factors that are derived from different species, both natural and recombinant. These growth factors have been used to promote long-term proliferation in cell culture and to induce cell differentiation in a variety of cell types under serum-free or reduced serum conditions.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage 2° to 8°C
Product Type Growth Factor
Format Natural
Formulation Lyophilized from ammonium acetate, carrier free, reconstitute in dH2O
For Use With (Application) A broad range of highly purified natural and recombinant lymphokine and growth factor biochemicals designed specifically for use in basic cell biology research
Purity or Quality Grade >98% by SDS-PAGE and pH 9.9 PAGE
Quantity 100 μg
Reactivity Mouse
Source Male Mouse Submaxillary Glands