Learn More
Corning™ DNA-BIND™ 96-Well Plates
GREENER_CHOICE
Product Code. 10525341 Shop All Corning Products
Click to view available options
Color:
Clear
White
Quantity:
10/Cs.
50/Cs.
Unit Size:
Pack of 10
Pack of 50
Product Code. 10525341
Brand: Corning™ 2505
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Convenient method for covalently immobilizing ssDNA by either the 5' or 3' end
  • Ready to use without surface preparation, prehybridization, or addition of expensive crosslinking reagents or highly purified oligos
  • Ideal for hybridization and amplification
  • Surface provides binding with high specificity and extremely low background
  • Flat-bottom wells
TRUSTED_SUSTAINABILITY
Specifications
Format Standard
Lid Without Lid
Array 8 x 12
Color Clear
Material Polystyrene
No. of Wells 96
Sterility Non-sterile
Well Shape Round
For Use With (Application) Ideal for hybridization and amplification
No. per Case 50/Cs.
No. per Pack 1/Pk.
Quantity 50/Cs.
Well Color Clear