Learn More
Corning™ Digital Hotplate and Stirrer Accessory Kit
GREENER_CHOICE
Product Code. 10451138 Shop All Corning Products
Click to view available options
Diameter (Metric):
0.8 cm
Unit Size:
Each
Product Code. 10451138
Brand: Corning™ 6795KIT
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
For Use With: PC-400D, PC-420D, PC-600D, PC-620D Digital Display Hotplates and Stirring Hotplates
This accessory kit is designed for all Corning digital display Hotplates and stirring Hotplates (Models PC-400D, PC-420D, PC-600D, PC-620D).
This Corning™ kit Cat. No. 6795KIT includes:
  • Cat. No. 6795PR, external temperature controller
  • Cat. No 440129, vertical support rod. Supplied as two 9 inch (22.9 cm) support rods that can be screwed together (18 inches)
  • Cat. No. 6970SR, stir bar retriever
Available for purchase separately and not included in the kit:
  • Cat. No. 440140, boss head clamp
  • Cat. No. 440141, holding rod
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) 0.8 cm
For Use With (Equipment) PC-400D, PC-420D, PC-600D, PC-620D Digital Display Hotplates and Stirring Hotplates
Includes Temperature Controller (6795PR), A Two Piece, 0.8 cm Diameter, 45.7 cm L Support Rod (440129) and Magnetic Stir Bar Retriever (6970SR)
Length (Metric) 45.7 cm
Type Hot Plate Kit