Learn More
Corning™ DeckWorks™ Bulk Pipet Tips
Shop All Corning Products
Click to view available options
Volume (Metric):
0.1 to 10 μL
1 to 200 μL
100 to 2000 μL
Color:
Natural
Yellow
Unit Size:
Pack of 1000
Pack of 768
Description
Corning DeckWorks bulk pipet tips are compatible with DeckWorks racks and packaged in resalable polyethylene bags. Tip sizes include 10 μL, 200 μL, and 1000 μL and offer maximum savings.
Specifications
Specifications
| Volume (Metric) | 100 to 2000 μL |
| Sterility | Non-sterile |
| Color | Natural |
| For Use With (Application) | Help reduce the production and waste of plastic materials |
| Tip Style | Round |
| Disposable | Single-use |
| For Use With (Equipment) | DeckWorks Racks |
| Graduated | Yes |
| Product Line | DeckWorks |
| Material | Polypropylene |