Learn More
Corning™ CrystalEX™ 96-Well Protein Crystallization Microplates
GREENER_CHOICE
Product Code. 10039661 Shop All Corning Products
Click to view available options
Surface Treatment:
High Binding
Non-treated
Unit Size:
Pack of 50
Product Code. 10039661
Brand: Corning™ 3773
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Designed for sitting-drop vapor diffusion high throughput protein crystallization
96-well Microplates
  • Features 96 large reservoir (reagent) wells and 96 corresponding protein wells
  • Conical bottom protein wells allow improved centering of the protein drop.
  • Compatible with manual pipettors and automation
  • Novel merged well design provides efficient vapor space for protein crystallization.
  • Reservoir and protein wells are positioned to be compatible with multi-head dispensing equipment.
384-well Microplate
  • Meets ANSI/SBS standard footprint dimensions for microplates
  • Ideal for fully automated crystal screening
  • Features 192 reservoir wells and 192 corresponding protein wells
  • Flat bottom protein wells are optimized for imaging of crystals.
  • Reservoir and protein wells are positioned to be compatible with multi-head dispensing equipment.
TRUSTED_SUSTAINABILITY
Specifications
Format Standard
Lid Without Lid
Array 8 x 12
Color Clear
No. of Wells 96
Sterility Non-sterile
Surface Treatment Non-treated
Well Shape Round
Includes 96 flat-bottom reservoir wells and 96 conical-bottom protein wells
No. per Case 50/Cs.
No. per Pack 10/Pk.
Quantity 50/Cs.
Depth (Metric) 3.1 mm