Learn More
Corning™ Costar™ Transtar™-96 96-Well Liquid Transfer System Accessories: Reservoir Liners
For use with 96-well plates
200.05€ - 226.82€
Specifications
Sterility | Sterile |
---|---|
For Use With (Application) | Ideal for repetitively filling multi-channel pipettors |
No. per Case | 100/Cs. |
No. per Pack | 1/Pk. |
Product Code | Brand | Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Price | Quantity & Availability | |||||
10143811
|
Corning™
4876 |
Open Reservoir Liner |
200.05€
Pack of 100 |
Estimated Shipment: 02-10-2024
Log in to see stock. |
|||||
10407611
|
Corning™
4878 |
8 Channel Reservoir |
220.18€
Pack of 100 |
In Stock
Log in to see stock. |
|||||
10311441
|
Corning™
4877 |
12 Channel Reservoir |
226.82€
Pack of 100 |
|
|||||
Description
For use with 96-well plates
These are sterile replacement open reservoir liners for the Transtar-96™ Well Liquid Transfer system.Specifications
Sterile | |
Ideal for repetitively filling multi-channel pipettors | |
100/Cs. | |
1/Pk. |