Learn More
Corning™ CellStripper Dissociation Reagent
BIOPHARMA
Product Code. 15313661
Change view
Click to view available options
Quantity:
6 x 100 mL
Unit Size:
Pack of 6
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15313661 6 x 100 mL Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15313661 Supplier Corning™ Supplier No. 25056CI
  • ONLINE EXCLUSIVE PRICE
    128.54€
    115.34€ / Pack of 6
    Save 13.20€
    10% Off
 Restricted Item
In Stock
Non-enzymatic cell dissociation solution gently dislodges adherent cells in culture
Designed to be an alternative to trypsin when used in conjunction with serum-free or serum-containing media
  • Formulated with a proprietary mixture of chelators
  • Reduces the risk of cell damage associated with protein digestive enzymes
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage 2° to 8°C
Form Liquid
Product Type Cell Culture Dissociation Reagent
Sterility Sterile
Quantity 6 x 100 mL