Learn More
Corning™ IPTG, Powder
BIOPHARMA
Product Code. 15393721
Change view
Click to view available options
Quantity:
1000 mg
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15393721 1000 mg 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15393721 Supplier Corning™ Supplier No. 46102RF
  • ONLINE EXCLUSIVE PRICE
    57.20€
    33.07€ / 1g
    Save 24.13€
    42% Off
 Restricted Item
Estimated Shipment: 09-02-2027
Produced under highest industry standards
IPTG (isopropylthiogalactoside)
  • Inducer of lac operon in bacteria
  • Used with X-gal for detecting lac gene activity
  • For screening of clones containing gene of interest in lac operon
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) For screening clones when used with X-Gal to detect lac gene activity
Quantity 1000 mg