Learn More
Corning™ Tryptic Soy Broth, Animal-Free, Liquid
BIOPHARMA
Product Code. 15363721
Change view
Click to view available options
Quantity:
100 mL
20,000 mL
Unit Size:
100mL
20 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15363721 20,000 mL 20 L
15333591 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15363721 Supplier Corning™ Supplier No. 46060LB
  • ONLINE EXCLUSIVE PRICE
    1435.00€
    851.45€ / 20 L
    Save 583.55€
    40% Off
 Restricted Item
Estimated Shipment: 26-11-2026
For an accurate assessment of sterility throughout the manufacturing process
Tryptic Soy Broth is a sterile-filtered, ready-to-use solution for use in sterility testing and validation in the manufacture of a liquid product. The use of this medium in place of the product intended for production allows for the assessment of the capability of an aseptic manufacturing process. Due to the rich nutrient base, TSB may be used in the cultivation of fastidious micro-organisms. This growth medium has animal-free components, making it ideal for testing the sterility of animal-free biopharmaceutical processes.
Order Info
Shipped under ambient conditions
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance USP
Content And Storage 15°C to 30°C
Form Liquid
For Use With (Application) For testing the sterility of animal-free biopharmaceutical processes
Quantity 20,000 mL