Learn More
Corning™ Metallocene Collection Container 2D Pillow Configuration
Shop All Corning Products
Click to view available options
Capacity (Metric):
50 L
100 L
Unit Size:
Each
Description
Corning collection bags are available in 2D hanging configurations with multiple bag volumes and connector configurations.
Specifications
Specifications
| Material | Metallocene |
| Sterility | Sterile |
| Capacity (Metric) | 100 L |
| Includes | Port 1, 2; 24 in. length tubing w/0.375 in. I.D. Male CPC conn. w/0.375 in. barb, dust cover and clamps; Port 3; 36 in. tubing 0.125 in. I.D. w/0.125 in. barb and male luer |
| No. per Case | 1 Ea. |
| No. per Pack | 1/Pk. |