Learn More
Corning™ Metallocene Collection Container 2D Pillow Configuration
Product Code. 17203574 Shop All Corning Products
Click to view available options
Capacity (Metric):
50 L
100 L
Unit Size:
Each
Product Code. 17203574
Brand: Corning™ 9110030
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Corning collection bags are available in 2D hanging configurations with multiple bag volumes and connector configurations.
Corning collection bags are available in 2D hanging configurations with multiple bag volumes and connector configurations.
TRUSTED_SUSTAINABILITY
Specifications
Material Metallocene
Sterility Sterile
Capacity (Metric) 50 L
Includes Port 1, 2; 24 in. length tubing w/0.375 in. I.D. Male CPC conn. w/0.375 in. barb, dust cover and clamps; Port 3; 36 in. tubing 0.125 in. I.D. w/0.125 in. barb and male luer
No. per Case 1 Ea.
No. per Pack 1/Pk.