Learn More
Corning™ Collection Bags
Shop All Corning Products
Click to view available options
Capacity (Metric):
0.5 L
1 L
5 L
10 L
20 L
50 L
100 L
200 L
Film Description:
EVA
ULDPE
Unit Size:
Each
Description
Corning collection bags are available in 2D hanging configurations with multiple bag volumes and connector configurations.
Specifications
Specifications
| Capacity (Metric) | 5 L |
| Tubing Style | Heat sealable flexible tubing |
| Film Description | EVA |
| No. per Case | 1/Ea. |
| No. per Pack | 1/Pk. |
| Port Detail | Port 1 - 0.37 in. Connector, 12 L x 0.37 I.D. x 0.62 in. O.D., chemically resistant, heat sealable flexible tubing, port 2 - male MPC connector with 0.37 in. barb, female MPC cap, port- 3 female MPC connector with 0.37 in. barb, male MPC plug, break-off i |
| Quantity | Each |