Description
Corning extracellular matrices (ECMs) enable researchers to mimic in vivo environments for 2D and 3D cell culture applications.
Specifications
Specifications
| Content And Storage | -20°C |
| For Use With (Equipment) | Falcon FluoroBlok™ Cell culture inserts and insert systems to label cells |
| Quantity | 1 mg |
For research use only.