Learn More
Corning™ Black and White 96-Well Polypropylene Assay Plates
Product Code. 17710698 Shop All Corning Products
Change view
Click to view available options
Color:
Black
White
Unit Size:
Pack of 100
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
17710698 White Pack of 100
10285421 Black Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17710698 Supplier Corning™ Supplier No. 3355
  • ONLINE EXCLUSIVE PRICE
    441.00€
    411.63€ / Pack of 100
    Save 29.37€
    6% Off
Estimated Shipment: 05-01-2027
Add to basket
Add to basket
Bulk Quote Request
Lower compound sticking and higher solvent resistance than polystyrene plates
  • Ideal for fluorescence and luminescence assays and for compound storage
  • Round-bottom wells
  • Untreated surface
TRUSTED_SUSTAINABILITY
Specifications
Format Standard
Array 8 x 12
Color White
Material Polypropylene
No. of Wells 96
Volume (Metric) Working 75 to 200 μL
Sterility Non-sterile
Surface Treatment Untreated
Well Shape Round
For Use With (Application) Ideal for fluorescence and luminescence assays and for compound storage