Learn More
Corning™ BioCoat™ Poly-L-Lysine Culture Dishes
Click to view available options
Diameter (Metric):
35 mm
60 mm
No. per Pack:
20/pk.
5/pk
No. per Case:
100/cs.
20/cs.
Unit Size:
Pack of 100
Pack of 20
Description
Suitable for promoting adhesion of transfected cell lines and other fastidious cell types. Designed to closely mimic in vivo environments and enhance expression of cell-specific morphology.
- Minimize assay failure due to cell detachment
- Matrix components are applied as an even, optically clear coating
- Precoated with Poly-L-Lysine to eliminate time-consuming preparation
- Lot-to-lot consistency provides reproducible experiments
- All lots tested and found negative for bacteria and fungi
- Cellware is tested for the ability to promote firm attachment of rat cerebellar granule cells
For adhesion, growth and differentiation of neuronal and transfected cell lines
Order Info
Bulk quantities available; please contact your Customer Service Representative.
Specifications
Specifications
| Bottom Shape | Round |
| Culture Area | 21.3 sq. cm |
| Shape | Round |
| Surface Treatment | Poly-L-Lysine |
| Type | Cell Culture Dish |
| No. per Case | 20/cs. |
| No. per Pack | 20/pk. |
| Diameter (Metric) | 60 mm |