Learn More
Corning™ BioCoat™ Poly-D-Lysine Culture Dish
GREENER_CHOICE
Product Code. 10399320
Click to view available options
Diameter (Metric):
35 mm
60 mm
100 mm
150 mm
Quantity:
10/cs.
100/cs.
20/cs.
40/cs.
5/cs.
Unit Size:
Pack of 10
Pack of 100
Pack of 20
Pack of 40
Pack of 5
Product Code. 10399320
Brand: Corning™ 354467
Ideal for applications involving adhesion, growth and differentiation of neuronal and transfected cell lines
  • All lots tested and found negative for bacteria and fungi
  • Cellware is tested for the ability to promote firm attachment of rat cerebellar granule cells
  • Coated with 0.2 micron filtered PDL in a cleanroom
Note: Dishes are not terminally sterilized and not assigned an SAL level. 
Order Info
Bulk quantities available; please contact your Customer Service Representative.
TRUSTED_SUSTAINABILITY
Specifications
Closure Color Clear
Closure Included Closure Included
Closure Material Polystyrene
Bottom Shape Round
Color Clear
Culture Area 9.6 sq. cm
Material Polystyrene
Shape Round
Surface Treatment Poly-D-Lysine
Type Cell Culture Dish
Includes Lid
Quantity 20/cs.
Diameter (Metric) 35 mm