Learn More
Corning™ BioCoat™ Angiogenesis System for Endothelial Cell Tube Formation
GREENER_CHOICE
Product Code. 10244152
Click to view available options
Quantity:
1
5
Unit Size:
Each
Pack of 5
Product Code. 10244152
Brand: Corning™ 354150
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Includes
A Falcon™ 24-Multiwell Insert Plate and a non-tissue culture-treated 24-well receiver plate with lid
Evaluate and quantitate endothelial cell invasion using real-time fluorescence and non-destructive sample preparation
  • Highly reproducible assay system
  • Ideal for screening anti-angiogenic agents
  • Rapid image acquisition and data quantification
TRUSTED_SUSTAINABILITY
Specifications
Pore Size 3 μm
Type Microplate
Quantity 5
No. per Pack 5/Pk.
No. per Case 5/Cs.
Surface Coating BioCoat
Color Black, Clear
No. of Wells 96