Learn More
Corning™ Adapters for LSE™ Compact Centrifuges
For use with Corning LSE high speed microcentrifuge.
46.67€ - 125.42€
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
15292479
|
Corning™
480134 |
0.5/0.4 mL tubes |
46.67€
Pack of 6 |
Estimated Shipment: 23-05-2024
Log in to see stock. |
|||||
15202489
|
Corning™
480135 |
0.2 mL tubes |
46.67€
Pack of 6 |
Estimated Shipment: 23-05-2024
Log in to see stock. |
|||||
15703338
|
Corning™
480140 |
10/15 mL tubes in 50 mL rotor |
125.42€
Pack of 6 |
Estimated Shipment: 18-07-2024
Log in to see stock. |
|||||
11317691
|
Corning™
480142 |
5/7/10 mL tubes in 15 mL rotor |
116.79€
Each |
Estimated Shipment: 23-05-2024
Log in to see stock. |
|||||
11307691
|
Corning™
480140 |
10/15 mL tubes in 50 mL rotor |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Description
The individual adaptors for tubes are used in the Corning LSE High Speed Microcentrifuge.
Specifications
Adapter | |
6/Pk. |