Learn More
Corning™ 96-Well, Cell Culture-Treated, Flat-Bottom Microplate
Shop All Corning Products
Click to view available options
Color:
Clear
Unit Size:
Pack of 100
Description
- Cell culture microplates are sterile and nonpyrogenic.
- Lids available where indicated
Cat. nos. 3798 and 3898 processed to improve hydrophilicity for hemagglutination and similar assays.
Cat. Nos. 3585 and 3595 have special low evaporation lids.
Cat. No. 3841 is aseptically manufactured.
Specifications
Specifications
| Format | Standard |
| Lid | With Lid |
| Culture Area | 0.32 cm2 |
| Array | 8 x 12 |
| Packaging | 50/Pk., 100/Cs. |
| Binding Type | TC-Treated |
| Color | Clear |
| Product Type | Assay Plate |
| Material | Polystyrene |
| Volume (Metric) Well | 360 μL |
| Show More |