missing translation for 'onlineSavingsMsg'
Learn More

Corning™ 96-Well, Non-Binding, Flat-Bottom, Half-Area Microplate
GREENER_CHOICE

Product Code. 10215462 Shop All Corning Products
Click to view available options
Color:
White, Clear Bottom
Unit Size:
Pack of 100
This item is not returnable. View return policy

Product Code. 10215462

Brand: Corning™ 3884

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Color:White, Clear Bottom; Sterility:Non-sterile; Lid:Without Lid

  • Bottoms are 60% thinner than conventional polystyrene microplates, resulting in lower background fluorescence and enabling readings down to 340 nm.
  • Opaque walls prevent well-to-well cross-talk.
  • Optically clear flat bottom permits direct microscopic viewing.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Format Half-Area
    Lid Without Lid
    Array 8 x 12
    Color White, Clear Bottom
    For Use With (Equipment) Standard automated handling and detection systems
    Material Polystyrene
    No. of Wells 96
    Volume (Metric) Working 25 to 125 μL
    Sterility Non-sterile
    Surface Treatment Non-binding
    Well Shape Flat Bottom (F-well)
    For Use With (Application) Ideal for reducing protein and nucleic acid binding at low concentrations and increasing assay signal to noise
    No. per Case 100/Cs.
    No. per Pack 25/Pk.
    Quantity 100/Cs.
    Well Color Clear
    Show More Show Less
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.