Learn More
Corning™ 384-Well Clear Polystyrene Microplates
GREENER_CHOICE
Product Code. 10206051 Shop All Corning Products
Change view
Click to view available options
Color:
Clear
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
10206051 Clear Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10206051 Supplier Corning™ Supplier No. 3640
  • ONLINE EXCLUSIVE PRICE
    1285.00€
    1197.02€ / Pack of 100
    Save 87.98€
    6% Off
Estimated Shipment: 07-01-2027
Add to basket
Add to basket
Manufactured from clear medical grade polystyrene for strength and visibility. Corning™ 364-Well Clear Polystyrene Microplates present an excellent solution for cell culture and colorimetric (absorbance) assays. Use 364-well microplates for laboratory miniaturization and automation.
  • Total well volume of 112 μL; working well volume of 20 μL – 80 μL
  • Cell culture microplates are sterile and nonpyrogenic.
  • The 384-well Universal Optics NBS microplate is manufactured using an advanced polymer with high clarity and improved chemical resistant properties.
  • TRUSTED_SUSTAINABILITY
    Specifications
    Format Standard
    Lid Without Lid
    Description Nonbinding (NBS™) surface is a Corning proprietary treatment technology used on polystyrene microplates to create a nonionic hydrophilic surface (polyethylene oxide-like) that minimizes molecular interactions
    Array 16 x 24
    Color Clear
    Product Type 384-Well NBS™ Microplate
    Material Polystyrene
    No. of Wells 384
    Volume (Metric) Working 20 to 80 μL
    Sterility Non-sterile
    Surface Treatment Non-treated
    Total Volume (Metric) Well 112 μL
    Well Shape Flat Bottom (F-well)
    For Use With (Application) Reducing protein and nucleic acid binding at low concentrations and increasing assay signal to noise
    No. per Case 100/Cs.
    No. per Pack 25/Pk.
    Well Color Clear