Learn More
Corning™ 384-Well Low-Volume Solid Microplates
Product Code. 10699921 Shop All Corning Products
Click to view available options
Color:
Black
White
Surface Treatment:
Non-binding
Non-treated
Tissue Culture
Volume (Metric) Working:
2 to 20 μL
2 to 25 μL
5 to 40 μL
Barcode:
Includes Barcode
No Barcode
Unit Size:
Pack of 100
Pack of 50
Product Code. 10699921
Brand: Corning™ 3674
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Same low reagent volumes as 1536-well plates without the need for new instrumentation
Polystyrene plates feature a unique square-to-round well design with a conical-shaped bottom that provides easier access and imaging for superior assay performance.
  • Bottom reduces fluid and air entrapment, greater signal-to-noise ratio and reduced FP signal (mP) when working with very low volumes
  • Each well has a raised rim to reduce cross-contamination during assay and liquid-handling process
  • Choose black plates for fluorescence applications or white for luminescence applications
High-Binding Surface Plates
  • Ideal for immunoassays
  • Can bind medium (>10kDa) and large biomolecules that possess ionic groups and/or hydrophobic regions
  • Working volume: 2 to 20μL
TRUSTED_SUSTAINABILITY
Specifications
Barcode No Barcode
Format Standard
Lid Without Lid
Array 16 x 24
Color White
Material Polystyrene
Volume (Metric) Well 50 μL
No. of Wells 384
Volume (Metric) Working 5 to 40 μL
Sterility Non-sterile
Surface Treatment Non-treated
Well Shape Round
For Use With (Application) Provides easier access and imaging for superior assay performance, fluorescence applications, reduce cross-contamination during assay and liquid-handling process
Quantity 100/Cs.
Well Color White