Learn More
Corning™ 384-Well Low-Volume Solid Microplates
GREENER_CHOICE
Product Code. 10583234 Shop All Corning Products
Click to view available options
Color:
Black
White
Surface Treatment:
Non-binding
Non-treated
Tissue Culture
Volume (Metric) Working:
2 to 20 μL
2 to 25 μL
5 to 40 μL
Barcode:
Includes Barcode
No Barcode
Unit Size:
Pack of 100
Pack of 50
Product Code. 10583234
Brand: Corning™ 3824BC
Same low reagent volumes as 1536-well plates without the need for new instrumentation
Polystyrene plates feature a unique square-to-round well design with a conical-shaped bottom that provides easier access and imaging for superior assay performance.
  • Bottom reduces fluid and air entrapment, greater signal-to-noise ratio and reduced FP signal (mP) when working with very low volumes
  • Each well has a raised rim to reduce cross-contamination during assay and liquid-handling process
  • Choose black plates for fluorescence applications or white for luminescence applications
High-Binding Surface Plates
  • Ideal for immunoassays
  • Can bind medium (>10kDa) and large biomolecules that possess ionic groups and/or hydrophobic regions
  • Working volume: 2 to 20μL
TRUSTED_SUSTAINABILITY
Specifications
Barcode Includes Barcode
Format Standard
Array 16 x 24
Color White
Material Polystyrene
No. of Wells 384
Volume (Metric) Working 5 to 40 μL
Sterility Non-sterile
Surface Treatment Non-binding
Well Shape Flat Bottom (F-well)
For Use With (Application) Provides easier access and imaging for superior assay performance, fluorescence applications, reduce cross-contamination during assay and liquid-handling process, reducing protein and nucleic acid binding at low concentrations and increasing assay signal
Well Color White